VWR® Evaporating Dishes, PTFE
Fournisseur: VWR International
Certificats
A propos de cet article
Isostatically molded, FDA-approved pure PTFE evaporating dishes are chemically resistant and inert with a smooth internal finish.
- Usable to 260°C (500°F)
- Autoclavable
Customers Who Bought This Also Bought