SP Bel-Art Cryo Tower Storage Systems, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These polycarbonate storage towers are ideal for benchtop or incubator use, transportation, freezer, and general-purpose storage.
- Polycarbonate construction
- Small bench space footprint
- Autoclavable
A 15.2x15.2 cm (6x6") footprint saves valuable bench space.
Available in 4- and 8-level configurations, both towers accommodate Cryo-Safe™ Vial Storage Box 47751-740. Autoclavable at 121°C (250°F).
Customers Who Bought This Also Bought