VWR® Gooch Crucibles, Porcelain
Fournisseur: VWR International
Certificats
A propos de cet article
Chemical-porcelain gooch crucibles are used in total suspended solids determinations.
- Glazed interior and exterior
- Chemical-resistant
Glazed inside and out except outside bottom surface and rim. Perforated bottom. The Gooch Crucible with Wall Holes has two holes for suspension in an extraction apparatus. For use in temperatures up to 1150°C (2102°F).
Ordering information: Covers are sold separately.
Customers Who Bought This Also Bought