KIMBLE® KONTES® Inlet Adapters, with Hose Connection, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Vertical adapters with vacuum take-off and drip tip.
- Top outer [ST] joint
- Bottom inner [ST] joint
- Both joints are same size
- With hose connection and drip tip
The outer Standard Taper joint at the top and inner Standard Taper joint at the bottom are the same size.
Customers Who Bought This Also Bought