KIMBLE® KONTES® 105° Vacuum Take-Off Adapters, Standard Stem, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Adapters are bent at a 105° angle for use in vacuum distillation.
- 105° angle bend
- Top [ST] outer joint
- Bottom [ST] inner joint
- With hose connection and drip tip
Featuring [ST] top outer and lower inner joints with a hose connector and drip tip.
Customers Who Bought This Also Bought