KIMBLE® KONTES® Straight Adapter Tubes with [ST] Joint, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Straight adapter with [ST] inner joint at one end and rubber connector.
- Rubber connector with hole in top included
- Accommodates plain stem thermometers, gas inlet tubes, and more
- [ST] inner joint at one end
Rubber connector with 6.4 mm (¼") hole in the top attaches to opposite end of tube to allow use with plain stem thermometers, gas inlet tubes, and more. Each tube is supplied with a connector, which also may be purchased separately.
Customers Who Bought This Also Bought