SP Bel-Art Antibody Saver Tray, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Minimize the use of expensive antibodies during incubations by using an optimally sized tray.
- Stackable
- Polypropylene construction
- Attached lid protects against spillage
- Autoclavable
These polypropylene trays are available for popular midi and mini gels and include an attached lid to protect against spillage. Single lane trays are supplied as a group of five joined lanes that can be easily separated to single lane prior to use. Trays are stackable, reclosable, and reusable.
Customers Who Bought This Also Bought