Synthware Anti-Splash Adapters, Modified
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
Anti-splash adapters prevent possible contamination from evaporator.
- Two 4 mm drain holes
- Top outer [ST] joint
- Bottom inner [ST] joint
With two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Upper joint is outer [ST] and lower joint is inner [ST].
Customers Who Bought This Also Bought