Synthware Anti-Splash Adapters
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
Anti-splash adapters are designed to prevent possible contamination from a rotary evaporator.
- Top outer [ST] joint
- Bottom inner [ST] joint
The upper joint is an outer [ST] joint, while the lower joint is an inner [ST] joint.
Customers Who Bought This Also Bought