Drying Tube Adapters, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These U-shaped drying tube adapters are manufactured from borosilicate glass.
- U-shaped
- Inner joint on one end
- One flared open end
Adapters feature an inner joint on one end and flared open end for a No. 3 rubber stopper and 6.4mm diameter x 50.8mm length (¼ x 2") glass tube. The stopper/tube can be purchased separately.
Customers Who Bought This Also Bought