SP Wilmad-LabGlass Offset Adapters, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
These offset adapters are manufactured from borosilicate glass.
- [ST] outer joint on one end
- [ST] inner joint on the other end
- Both joints are same size
Adapters have a Standard Taper outer joint on one end and the same size Standard Taper inner joint on the other.
Customers Who Bought This Also Bought