Cleanroom Wiper Dispensers, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Wiper dispensers are made of clean, clear, 6 mm (1/4") thick PETG with flame-treated edges for a smooth finish.
- Protective covers help maintain the cleanliness of the wipers
- Table top only
Keep 22.9x22.9 cm (9x9") or 30.5x30.5 cm (12x12") wipers clean and easily accessible in these convenient wiper dispensers.
Customers Who Bought This Also Bought