Cleanroom Wiper Dispensers, Hinged Lid, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Dispensers are for use with 22.9x22.9cm (9x9") and 30.5x30.5cm (12x12") wipers
Protect cleanroom wipers from contamination while keeping them convenient. Hinged lid makes for easy loading and retrieval, and provides maximum protection when closed. Wipers are kept accessible through front opening. Made of clean, clear, 6mm (1/4") thick acrylic or PETG with rubber pads on bottom corners to prevent slipping. The PETG material is safe with IPA. Edges are flame-treated for a smooth finish. Durable full-length hinge. Free-standing or wall-mountable.
Customers Who Bought This Also Bought