Accessories for Spill Containment Pallet/Platforms with Grating, Eagle Mfg
# de catalogue 56610-212
Fournisseur: Eagle Manufacturing
Spécifications
- Description:Ramp
- Cat. no.:56610-212
- L×W×H:115.5 x 81.2 x 20.3 cm (451/2 x 32 x 8")
- Load Capacity:680 kg (1500 lbs.)
- Weight:14.5 (32)
- Supplier No.:1689