Synthware Distillation Head Adapter, Kemtech America
# de catalogue 60000-642
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware Distillation Head Adapter
- Cat. no.:60000-642
- Supplier No.:A542440
Spécifications
A propos de cet article
Synthware distillation head adapter is vacuum jacketed.
- [ST] 10/18 top joint
- Accommodates thermometer
- Thermometer not included
The [ST] 10/18 top joint accommodates a 76mm immersion thermometer. Other [ST] joints are 24/40.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.
Customers Who Bought This Also Bought