Nalgene® Pans, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Pans are lightweight, durable, chemically inert, and autoclavable
For repeated autoclaving of small instruments and labware. Can also be used for soaking, cleaning, storing, or carrying general lab products. Heavy-duty construction with extra strong rims. Tapered for nesting.
Customers Who Bought This Also Bought