Synthware Anti-Splash Adapters with Fritted Disc, Modified, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Anti-splash adapters prevent possible contamination from evaporator.
- Two 4mm drain holes
- Extra coarse fritted disc
Adapters feature two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask and an extra coarse fritted disc sealed above lower inner joint. Frit reduces foam overflow from being drawn into the condenser.
Customers Who Bought This Also Bought