ContactUsLinkComponent
United Scientific™ Advanced Electricity And Magnetism Kit
United Scientific™ Advanced Electricity And Magnetism Kit
# de catalogue 470356-044
New Product
KIT ADV ELECTRICITY AND MAGNETISM
United Scientific™ Advanced Electricity And Magnetism Kit
# de catalogue 470356-044
New Product
Spécifications
  • Description:
    Advanced electricity and magnetism kit
Spécifications
A propos de cet article
Explore several aspects of electricity and magnetism with this hand on kit.
  • 25 activities addressing seven key topics
  • Appropriate for elementary to high school
  • Aligned with NGSS Standards
  • Includes detailed activity guide
This comprehensive 47-piece kit introduces students to the basic principles of electricity and magnetism. Includes materials for studying static electric charges, magnetic attraction and repulsion, magnetic fields, electrical circuitry, voltage, current and resistance, and series and parallel connection.
Electromagnetism topics include the force in a magnetic field and electric motor and generator basics. Light emitting diodes, a solar cell, a reed switch, a buzzer, and an electric bell are provided for further investigations. The components are mounted on sturdy molded bases and connections are made with reliable spring terminals. An activity guide details the experiments.
Ordering information: Requires three AA batteries (not included).
Delivery: Kit includes DC ammeter (1); DC voltmeter (1); Battery holders, AA size (3); Knife switches, single pole, single throw (2); Knife switch, single pole, double throw (1); Lamp holders (3); Miniature incandescent lamps (8); Resistor board (5, 10, 15 Ω, 'Unknown') (1); Slide Rheostat (20 Ω, 2 A) (1); Wire resistance board (1); Conductivity test board (1); Bar magnet (1); U-shaped magnet (1); Ring magnets (3); Ring magnet carriage (1); Compass (1); Magnetic field visualizer (1); Electric bell (1); Electromagnet (1); Electromagnetic force device (1); Motor model (1); DC miniature motor (1); Hand generator (1); LED board with two LEDs (1); Solar cell (1); Buzzer (1); Magnetic reed switch (1); Set of electrostatic test objects (1); Set of 16 connecting cords (1); Set of electrodes and spares (1).
Customers Who Bought This Also Bought