Grain of Wheat Bulbs
Fournisseur: American Scientific
New Product
Certificats
A propos de cet article
Flexible and easy alternative to standard mini bulbs: no holder required!
- Comes with a teacher’s guide
- Sold in packs of 100
- Incandescent bulbs take a maximum of 12 V and 500 mA
- LED bulbs take a maxiumum of 3 V and 3 mA
This is a new generation of ultra-bright bulbs offering easy replacement, making them ideal for use in a variety of miniature equipment and circuit applications. The wheat bulbs can also be used in control panels and high-quality commercial and industrial controls. They are similar to the wire-ended lamps but they have stranded wire leads about 8" (200 mm) long. The bulb is about 0.13" (3 mm) dia. and 0.25" (6 mm) long - the size of a grain of wheat.
Or, upgrade your outdated bulbs to the 21st century! These white color LED bulbs offer an easy replacement, making them ideal for use in a variety of miniature equipment and circuit applications.
Customers Who Bought This Also Bought