A propos de cet article
Versatile indoor or outdoor system which can be used for Aquaponics, Hydroponics, or Water Garden.
- Rated for indoor and outdoor use
- Durable, weatherproof grow bed and fountain reservoir
- Includes a submersible 160 gph pump
- Control the flow rate with customizable plumbing fittings
- Room for 6 or more plants
Add a touch of beauty and a splash of life to any room with the AquaSprouts Fountain! Water feature, planter, and eye-catching decor, the Fountain can be used as an aquaponic or hydroponic system, or a simple water garden.
Similar to AquaSprouts Gardens, the expanded clay grow media replaces soil from traditional gardening and serves as the support for your plants.
Use any small freshwater fish to skip the added fertilizer and let Mother Nature be your guide. The fish provide nutrients for the plants and the plants clean the water for the fish. Once established this means a low maintenance ecosystem at your fingertips.
Not in the mood for fish? Use water-soluble nutrients to keep your plants happy and healthy with ease. Enjoy a tranquil water garden while growing your own personal oasis.