Fixed Blade Scalpels
Supplier: AFTAB AHMED SE
Certificates
About this item
More durable than replaceable blades.
- Pre-sharpened
Fixed blade scalpels are more durable than replaceable blades. With less flex there is less likelihood of blade breakage during a dissection.
Best used for more coarse dissection near hard tissue, these offer three different materials of sharpening.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought