Synthware Distillation Adapter, Bent 105°, Kemtech America
# de catalogue 60000-370
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:24/40
- Cat. no.:60000-370
- Supplier No.:A412440
Spécifications
A propos de cet article
Synthware 105° bent distillation adapter.
- Multiple uses
- 105° angle bend
This adapter can also be used as a rotary evaporator washer.
Customers Who Bought This Also Bought