Synthware Inlet Adapters, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Use inlet adapters with gas inlet tubes or plain stem thermometers.
- Inner [ST] joint
- One-holed rubber adapter included
Features an inner [ST] joint. Supplied with one-holed rubber adapter.
Ordering information: For a replacement rubber adapter, contact your Avantor representative.
Customers Who Bought This Also Bought