Synthware Distilling Adapter, 105° Angle, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Synthware 105° angle bent distillation adapter.
- Outer [ST] joint at top
- Tapered drip tube
Features an outer [ST] joint at top. Tapered drip tube extends into open receivers.
Customers Who Bought This Also Bought