Mini Adapters, No. 7 Ace-Thred with PTFE Ferrule, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These mini adapters are suitable for most vacuum work.
- Inner joint at bottom
- No. 7 Ace-Thred at top
- Manufactured from borosilicate glass
Adapters feature an inner joint at bottom and No. 7 Ace-Thred at top.
Ordering information: Supplied with nylon bushing and PTFE ferrule in place of O-ring. For replacement ferrules, see 89075-734 series.
Customers Who Bought This Also Bought