Adapter, Offset, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Offest adapters are used to gain better vertical clearance when using addition funnels, condensers, or other apparatus.
- Bottom inner and top outer joints are slightly offset
- Made of borosilicate glass
Both bottom inner and top outer joints are slightly offset from each other. The 10° angled offset adapters are designed for use with reactor heads that have 10° angled side necks for transitioning to 90° vertical.
Customers Who Bought This Also Bought