VWR® Flexible Bellow Joints, PTFE
Fournisseur: VWR International
Certificats
A propos de cet article
Flexible bellows are constructed of pure, non-contaminating PTFE.
- Connect directly to standard ground glass joints
- Usable up to 260°C (500°F)
- Inert
Bellows correct misalignment and relieve stress in glassware assemblies. Transverse sealing rings provide medium to high vacuum performance and reduce the probability of glass bursting during temperature cycling.
Customers Who Bought This Also Bought