VWR® Sound Level Meter
# de catalogue 12777-836
Fournisseur: VWR International
Spécifications
- Description:Traceable® sound level meter
- Weight:269 g (9.5 oz.)
Spécifications
A propos de cet article
This Traceable® sound level meter features three decibel ranges for greater precision: low (30 - 80 dB), medium (50 - 100 dB), and high (80 - 130 dB).
- Adjustable esponse time on the meter may be set to fast (0.2 s) or slow (1.5 s)
- A memory data-hold function stores the highest reading achieved for later display
- Meter uses a 1.3 cm (0.5") condenser microphone, and a recorder jack captures readings (AC and DC 10mV/dB)
- Provides weightings for both A (frequencies for 500 to 10kHz) and C (frequencies from 30 to 10kHz) to meet IEC 651 Type-2 standards
Resolution: 0.1 dB
Accuracy: ±2 dB
W×L×H: 7.6×25.4×2.5 cm (3×10×1")
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Meter is supplied with a 9V alkaline battery and carrying case.
Customers Who Bought This Also Bought