KIMBLE® KONTES® 75° Distilling Adapter, DWK Life Sciences
# de catalogue KT158000-2440
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Width:140 mm
- Height:130 mm
- Supplier No.:158000-2440
Spécifications
A propos de cet article
KONTES® 75° bent distillation adapter.
- [ST] 24/40 inner joints
- 75° angle bend
Features [ST] 24/40 inner joints at each end.
Customers Who Bought This Also Bought