KIMBLE® KONTES® Distilling Adapters, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Distilling adapter is useful in the sub-boiling separation of mixtures with volatile components.
- Parallel [ST] joints
- Adapter T169500-2440 has parallel [ST] joints
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought