KIMBLE® KONTES® AIRLESS-WARE® Gas Inlet Adapter with Glass Plug, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Gas inlet type adapters with inner [ST] joint.
- Inner [ST] joint
- Includes glass and PTFE plug
- Manufactured from 33 expansion, low extractable borosilicate glass
For use with a sleeved stopper.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with KT801001-0002 glass plug and patented pressure/vacuum retainer and KT882311-0000 sleeved stopper.
Customers Who Bought This Also Bought