Synthware Claisen Adapters, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
The three inlets of this adapter allow for multiple entries into a single neck flask.
- Claisen style adapters
- Three inlets
One side joint is set at a 45° angle.
Customers Who Bought This Also Bought