Synthware Connecting Adapters with Three Arms, Two Inlet, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
The three-arm design of this adapter allows for the assembly of three vessels.
- Three arms
- Two inlets
Connecting adapter with two female joints on top and a male joint below.
Customers Who Bought This Also Bought