Synthware Distillation Adapters, Kemtech America
# de catalogue 60000-656
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:24/40
- Cat. no.:60000-656
- Supplier No.:A362410
Spécifications
A propos de cet article
Distillation adapter accommodates a 76 mm immersion thermometer.
- Accommodates thermometer
- Thermometer not included
Features a [ST] 10/18 joint on the vertical side tube for a 76 mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.
Customers Who Bought This Also Bought