Synthware Distillation Connecting Adapters, Vigreux, Kemtech America
# de catalogue 60000-644
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware distillation connecting adapters
- Cat. no.:60000-644
- Supplier No.:A131024
Spécifications
A propos de cet article
Distillation connecting adapter accommodates a 76 mm immersion thermometer.
- Accommodates thermometer
- Thermometer not included
Top [ST] joint is 10/18. Side [ST] joint is 24/40. Other [ST] joint is 24/40.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.
Customers Who Bought This Also Bought