PIG® All-Purpose Wipers, New Pig
Fournisseur: New Pig
Certificats
A propos de cet article
Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping.
Excellent for a variety of wiping applications from quick cleanups to degreasing.
Available 1/4 folded and poly wrapped for easy handling and loading. Convenient pop-up box lets you take just one wiper at a time while keeping remaining stock clean and dry. Center-pull roll fits wall-mounted unit and dispenses one at a time to help prevent overuse.
Made thick and absorbent to quickly soak up oils, degreasers and other liquids. Low-lint material won't leave residue or create a mess.
Customers Who Bought This Also Bought