Synthware Claisen Adapter, Kemtech America
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
This Claisen-style adapter provides dual entry into a variety of flasks.
- Allows dual entry
- Use with a variety of flasks
The upper joints are outer [ST] and the bottom joint is an inner [ST].
Customers Who Bought This Also Bought