Septum Inlet Adapters with Stopcock, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Sampling adapters are designed for use in handling air-sensitive materials.
- Bottom [ST] joint
- Manufactured from borosilicate glass
- Two septa included
Adapters feature a [ST] joint at bottom, 2mm bore stopcock, and two right angle septum ports at top.
Ordering information: Supplied with two septa. For replacement septa, contact your Avantor sales representative.
Customers Who Bought This Also Bought