PYREX® Three-Way Connecting Tube with Parallel Sidearm and [ST] Joints, Corning
# de catalogue 89091-552
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Description:Claisen Adapter, 160 mm
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89091-552
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:9040-24
Spécifications
A propos de cet article
This Claisen type three-way connecting tube features a parallel sidearm.
- Claisen-style adapter
- Parallel sidearm
- All joints are [ST] 24/40
Featuring [ST] outer joints at upper end and on parallel sidearm. [ST] inner joint on lower end. All joints on each tube are [ST] 24/40.
Customers Who Bought This Also Bought