Alliance Contamination Control Frocks, HPK Industries
Fournisseur: HPK Industries
Certificats
A propos de cet article
Smooth outer fabric surface lowers particle cling and build-up in cleanroom applications
Built-in “cooling system” dissipates heat and moisture for superior worker comfort. Snap front, elastic wrists, no pocket.
Bound seam construction. Frock material acts as a liquidproof barrier for protection from liquids used in biotechnology and pharmaceutical cleanrooms. Unique antistatic system assures static dissipation in sensitive cleanroom work areas.
Ordering information: Individually packaged in a sealed bag.
Spécifications
- Catalog No:
- 10832-240
- 10832-242
- 10832-166
- 10832-168
- 10832-234
- 10832-236
- 10832-238
- 10832-170
- 10832-172
- 10832-174
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory