DuPont™ ProClean® Frocks, Front Snap Closure
Fournisseur: DuPont
Certificats
A propos de cet article
ProClean® offers improved non-hazardous light liquid splash protection.
- Serged seams
- Bound neck
- Set sleeve design
- Covered elastic wrists
- Front snap closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Spécifications
- Catalog No:
- CA13452-504
- CA89129-702
- CA13452-502
- CA13452-512
- CA13452-514
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Spécifications
Informations connexes