ContactUsLinkComponent
Cotton Diaper Towels, Lymtech®, FG Cleanwipes
Cotton Diaper Towels, Lymtech®, FG Cleanwipes
# de catalogue 82001-038
Fournisseur:  Lymtech
Cotton Diaper Towels
Cotton Diaper Towels, Lymtech®, FG Cleanwipes
# de catalogue 82001-038
Fournisseur:  Lymtech
Spécifications
  • W×L:
    45.7 x 45.7 cm (18 x 18")
  • Cat. no.:
    82001-038
  • Supplier No.:
    DNB1BOE-HH
Spécifications
A propos de cet article
Pure cotton diaper cloth towels are strong, soft, and absorbent.
Towels are ideal for wiping, polishing, and general hygiene requirements. Durable towels feature a hemmed edge to allow for re-washing and re-use, minimizing the waste of disposables. Soft enough to use on sensitive surfaces.
Customers Who Bought This Also Bought