VWR® Cotton Pads
# de catalogue 21902-985
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- W×L:10.2 x 10.2 cm (4 x 4")
- Color:White
- Description:Cotton Pad
- Cat. no.:21902-985
Spécifications
A propos de cet article
Soft, pure, highly absorbent pads will not scratch the most delicate coatings or surfaces.
Economical, all-purpose 100% pure cotton pads. Useful for the printing industry. Soft, pure, highly absorbent pads will not scratch the most delicate coatings or surfaces. Will not contaminate developing solutions or chemicals.
- No Binders or chemicals
- Excellent wet strength
Economical, all-purpose 100% pure cotton pads. Useful for the printing industry. Will not contaminate developing solutions or chemicals.
Customers Who Bought This Also Bought