Bench and Daub Safety Can, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
For safer cleaning of small parts or swabbing larger ones
Seamless 24-gauge terne steel construction. Dasher screen acts as fire baffle. Components are easily removed for cleaning.
Certifications: FM approved. Meets OSHA requirements.
Ordering information: Safety can 21815-206 is supplied without lid.
Customers Who Bought This Also Bought