SP Bel-Art Safety Waste Jug, Bel-Art Products, a part of SP
# de catalogue 13197-284
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Cap Size:45 mm
- Capacity:20 L (5 gal.)
- Description:Polyethylene Waste Jug
- Cat. no.:13197-284
- W×D×H:30.5 x 24.4 x 40.6 cm (12 x 9⁵/₈ x 16")
- Supplier No.:119190000
Spécifications
A propos de cet article
For safe, in-plant handling and temporary storage of spent solvents or other liquid wastes awaiting proper disposal.
- Flared top on snap-on cap helps retain fumes
- Translucent
- HDPE jug construction
For easy filling, the screw-on top has a funnel-like flare, 12 cm (43/4") in diameter. A snap-on cap seals the flared top to retain fumes. Translucent, high-density graduated polyethylene jug.
Customers Who Bought This Also Bought