Nonmetallic Dispenser Cans, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
For easy one-hand dispensing of flammable liquids directly onto work
Dispensers 56609-744 and -748 are FM approved.
Brass dispenser valves are leakproof and self-closing to minimize spills and control excess. Rugged polyethylene body.
Customers Who Bought This Also Bought