Safety Cans with Faucet, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
For controlled dispensing of flammable liquids into small vessels using built-in, self-closing faucet valve.
Leave the can on the cabinet shelf or benchtop while you dispense solvents. Minimizes chance of spills or strain from lifting a heavy can. Helps meet OSHA standards. For use with alcohols, acetone, ether, toluene, xylene, and more.
Capacity: 18.9L (5gal.). Available in coated metal or Type 316 stainless steel with nitrile rubber gasket.
FM approved.
Customers Who Bought This Also Bought