Drum Faucet, Eagle Manufacturing
# de catalogue 56610-550
Fournisseur: Eagle Manufacturing
Spécifications
- Description:Brass Faucet
- Cat. no.:56610-550
- Supplier No.:1974-B
Spécifications
A propos de cet article
Self-closing brass faucet fits 1.9 cm (3/4") opening in drum
With internal flame arrester for safety.
Certifications: FM approved.
Customers Who Bought This Also Bought