Jumbo Stacking Cabinets, SciMatCo
Fournisseur: Avantor
Certificats
A propos de cet article
Jumbo Stacking Cabinets are constructed of 2.5 cm (1") thick, multi-ply, exterior-grade plywood and finished with easily identified, chemical-resistant, yellow or blue epoxy paint.
- All cabinets have a liquid-tight, 5.1 cm (2") trough including polypropylene liners to contain and easily clean up accidental spills
Available as an accessory, Polypropylene isolation compartment 18500-072 fits in units 18500-068 and 14212-950 and isolates nitric acid or nitric acid spills from all other products stored in these cabinets. Cabinets include suitable locking mechanism and keys. Flammables cabinet 14002-622 has one adjustable shelf. Acid;18500-068 and Corrosive;14212-950 cabinets have one fixed position shelf.
Designed to stack up to two units high, the cabinets can also be used as stand-alone units. Cabinets may be placed under any work surface with a 81.9cm (321/4") clearance.
Certifications: Cabinets meet all applicable NFPA and OSHA standards.
Ordering information: Flammables cabinets with self-close doors are also available; contact your Avantor Furniture Specialist for more information.
Customers Who Bought This Also Bought