Wooden Acid Cabinets, SciMatCo
Fournisseur: Scientific Materials
Certificats
A propos de cet article
All wood cabinets with no metal in the interior construction will never rust or corrode.
- Cabinets features: one fixed-position shelf
- Interlocking wooden door assembly
- Leveling feet and polypropylene lock assembly
- Cabinets are supplied with a padlock and two keys for security
Polypropylene nitric acid isolation compartment available as an accessory (18500-072) fits inside cabinets to isolate nitric acid or other incompatible chemicals stored within the same cabinet. A separate entirely polypropylene slide-in isolation compartment designed to isolate nitric acid or other incompatible chemicals stored within the same cabinet.
The 2.5 cm (1") thick high-density plywood is finished with epoxy paint, and “ACID” printed in 15.2 cm (6") high red letters. The recessed top includes a liquid-tight polypropylene tray and the floor has a 5.1 cm (2") trough to contain spillage.
Certifications: Meets or exceeds OSHA standards.
Customers Who Bought This Also Bought